Learn More
CERILLIANT™ Phendimetrazine tartrate tartrate salt Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore P1271ML
Description
An analytical reference standard suitable for use in LC/MS or GC/MS methods for clinical toxicology, forensic analysis, urine drug testing, or pharmaceutical research applications. Phendimetrazine, a prodrug of phenmetrazine, is a stimulant drug of the morpholine chemical class used as an appetite suppressant marketed under the names Bontril, Adipost, Anorex-SR, Appecon, Melfiat, Obezine, Phendiet, Plegine, Prelu-2, or Statobex.Specifications
| 67-56-1 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| 341.36 | |
| 1 mL |
| C12H17NO · C4H6O6 | |
| 1.0 mg/mL in methanol | |
| Ampule | |
| 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation