missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Phenazepam-D4 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001864
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001864 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001864 Supplier Merck Emd Millipore Supplier No. P0931ML

Certified reference material

A benzodiazepine developed in Russia, phenazepam is used in Europe for treatment of epilepsy, alcohol withdrawal syndrome, and insomnia. Phenazepam has gained popularity as a recreational drug and is sold under street names such as Zannie, Bonzai, Bonzai Supersleep, and Kronic. This Certified Spiking Solution™ is suitable for use as an internal standard for quantification of phenazepam in urine or other biological matrices by LC/MS or GC/MS.

Specifications

CAS 75-05-8, 1184980-42-4
Linear Formula C15H6D4N2OBrCl
UN Number UN 1648 3 / PGII
Concentration 100 μg/mL in acetonitrile
Formula Weight 353.63
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation