missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Pentaerythritol tetranitrate Solution, 10 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001338
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001338 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001338 Supplier Merck Emd Millipore Supplier No. ERP109S5ML
Only null left

Certified reference material

Specifications

CAS 75-05-8, 78-11-5
Linear Formula C5H8N4O12
MDL Number MFCD00058681
UN Number UN 3379 - DOT UN 1648 class 3 - PG 1 - Desensitized explosive, liquid, n.o.s., HI: all (not BR)
Concentration 10 mg/mL in acetonitrile
Synonym Pentaerythritol tetranitrate, PETN
Formula Weight 316.14
Packaging Ampule
Quantity 5 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation