missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ PCP-D5 (Phencyclidine-D5) Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001823
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001823 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001823 Supplier Merck Emd Millipore Supplier No. P0061ML

Certified reference material

Phencyclidine, commonly known as PCP, is a recreational drug with hallucinogenic and neurotoxic effects. With street names such as 'angel dust' or 'wet', PCP comes in either powder or liquid forms. Phencyclidine-D5 is a stable-labeled internal standard suitable for quantitation of PCP levels in urine, blood or hair by LC/MS or GC/MS for testing applications in urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods.

Specifications

CAS 67-56-1, 60124-86-9
Linear Formula C17H20ND5
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Synonym Phencyclidine-d5
Formula Weight 248.42
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation