missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ p-Fluorofentanyl Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001373
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001373 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001373 Supplier Merck Emd Millipore Supplier No. F0490.5ML

Certified reference material

p-Fluorofentanyl is an opioid analgesic being an analogue of fentanyl developed by Janssen Pharmaceutica in the 1960s. p-Fluorofentanyl potently activates the human opioid receptor and is abused recreationally. A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls for p-Fluorofentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing or forensic analysis applications.

Specifications

Packaging Ampule
CAS 90736-23-5
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol
Linear Formula C22H27FN2O
MDL Number MFCD06411158
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 0.5 mL
Formula Weight 354.46

CERILLIANT is a registered trademark of Cerilliant Corporation