missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Oxymorphone-D3 Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001807
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001807 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001807 Supplier Merck Emd Millipore Supplier No. O0191ML

Certified reference material

This internal standard is suited for quantification of oxymorphone levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods. Oxymorphone, sold under trade names such as Opana™, Numorphan™, and Numorphone, is an opiate analgesic indicated for the relief of moderate to severe pain.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 1.0 mg/mL in methanol
Linear Formula C17H16NO4D3
MDL Number MFCD00673302
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 304.36

CERILLIANT is a registered trademark of Cerilliant Corporation