missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Oxcarbazepine Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Description
Oxcarbazepine, marketed under the trade name Trileptal™, is an anticonvulsant developed and prescribed for treatment of epilepsy. In recent years, Oxcarbazepine has shown efficacy in treatment of mood disorders. This certified solution standard is suitable as starting material for the preparation of calibrators and controls in oxcarbazepine testing by GC/MS or LC-MS/MS.
Specifications
Specifications
| CAS | 75-05-8, 28721-07-5 |
| Linear Formula | C15H12N2O2 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 1.0 mg/mL in acetonitrile |
| Formula Weight | 252.27 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation