missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Oxcarbazepine-13C6 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001816
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001816 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001816 Supplier Merck Emd Millipore Supplier No. O0391ML
Only null left

Certified reference material

This stable-labeled internal standard is suitable for use in quantitation of oxcarbazepine levels or isotope dilution methods by GC/MS or LC/MS for applications in pain prescription monitoring, clinical toxicology, therapeutic drug monitoring or urine drug testing. Oxcarbazepine, marketed under the trade name Trileptal™, is an anticonvulsant developed and prescribed for treatment of epilepsy. In recent years, Oxcarbazepine has shown efficacy in treatment of mood disorders.

Specifications

CAS 75-05-8
Linear Formula 13C6C9H12N2O2
MDL Number MFCD00865307
UN Number UN 1648 3 / PGII
Concentration 100 μg/mL in acetonitrile
Formula Weight 258.22
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation