missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Ocfentanil Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001818
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001818 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001818 Supplier Merck Emd Millipore Supplier No. O0470.5ML

Certified reference material

A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls for ocfentanil LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, prescription monitoring, or forensic analysis applications. Ocfentanil, also known as A-3217, is a potent synthetic opioid analgesic drug structurally related to fentanyl. It is used in combination with other opioids and analgesics to induce or supplement general anesthesia.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol
Linear Formula C22H27FN2O2
MDL Number MFCD00864238
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 0.5 mL
Formula Weight 370.46

CERILLIANT is a registered trademark of Cerilliant Corporation