missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Nortilidine Hydrochloride, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Description
Nortilidine is the primary, active metabolite of tilidine, a synthetic opioid analgesic used mainly in Germany, Switzerland and Belgium for treatment of moderate to severe pain, both acute and chronic. This new Snap-N-Spike™ solution standard is suitable for numerous GC/MS and LC/MS applications from clinical toxicology and forensic analysis to calibrator preparation and urine drug testing.
Specifications
Specifications
| CAS | 67-56-1, 34596-11-7 |
| Linear Formula | C16H21NO2 · HCl |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Concentration | 1.0 mg/mL in methanol (as free base) |
| Synonym | Nortilidine hydrochloride solution |
| Formula Weight | 295.80 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation