missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Norfentanyl-D5 oxalate Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001709
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001709 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001709 Supplier Merck Emd Millipore Supplier No. N0301ML

Certified reference material

An internal standard suitable for norfentanyl testing of urine, blood, or hair samples or isotope dilution methods by GC/MS or LC/MS in applications such as pain prescription monitoring, clinical toxicology, urine drug testing, or forensic analysis. Norfentanyl is a major urinary metabolite of fentanyl, a potent synthetic analgesic sold under various trade names including Sublimaze, Duragesic™, Actiq™, and Fentora™.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol (as free base)
Linear Formula C16D5H17N2O5
MDL Number MFCD08272800
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 327.39

CERILLIANT is a registered trademark of Cerilliant Corporation