missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Norcocaine Hydrochloride Solution, 1.0 mg/mL in Acetonitrile (as free base), Sold by MilliporeSigma™ Supelco™
Description
Norcocaine is a primary urinary metabolite of the popular recreation drug cocaine. Cocaine is a central nervous system stimulant with appetite suppressant and topical anesthetic properties. This certified solution standard is applicable for use in urine drug testing, clinical toxicology or forensic toxicology applications by GC/MS or LC/MS.
Specifications
Specifications
| CAS | 75-05-8, 61585-22-6 |
| Linear Formula | C16H19NO4 · HCl |
| MDL Number | MFCD00242703 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 1.0 mg/mL in acetonitrile (as free base) |
| Formula Weight | 325.79 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation