missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Norcarfentanil oxalate oxalate salt Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001767
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001767 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001767 Supplier Merck Emd Millipore Supplier No. N1141EA

Certified reference material

A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls for norcarfentanil LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, prescription monitoring, or forensic analysis applications. Norcarfentanil is a major urinary metabolite of carfentanyl, a potent synthetic opioid analgesic drug structurally related to fentanyl. It is used in combination with other opioids and analgesics to induce or supplement general anesthesia.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol (as a free base)
Linear Formula C16H22N2O3 · C2H2O4
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 0.5 mL
Formula Weight 380.39

CERILLIANT is a registered trademark of Cerilliant Corporation