missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Norbuprenorphine Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001791
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001791 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001791 Supplier Merck Emd Millipore Supplier No. N9121ML

Certified reference material

Norbuprenorphine is the primary active metabolite of the opiate buprenorphine in urine. This Certified Spiking Solution™ is suitable as a starting material for use as calibrators and controls in GC/MS and LC/MS methods in forensic analysis, clinical toxicology, urine drug testing, or pain prescription monitoring. Buprenorphine, sold under trade names such as Buprenex™, Subutex™ and Suboxone™, is an opiate drug used to treat opioid addiction and to moderate acute pain.

Specifications

Packaging Ampule
CAS 78715-23-8
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol
Linear Formula C25H35NO4
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 413.55

CERILLIANT is a registered trademark of Cerilliant Corporation