missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Norbuprenorphine Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001731
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001731 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001731 Supplier Merck Emd Millipore Supplier No. N0591ML

Certified reference material

A new Certified Spiking Solution™ suitable for use in clinical toxicology, forensic testing, or pain prescription monitoring.

Specifications

CAS 67-56-1, 78715-23-8
Linear Formula C25H35NO4
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 413.55
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation