missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Norbuprenorphine-D3 Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001795
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001795 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001795 Supplier Merck Emd Millipore Supplier No. N9211ML
Only null left

Certified reference material

Norbuprenorphine is the primary active metabolite of the opiate buprenorphine in urine. This internal standard is suitable for quantitation of norbuprenorphine levels or isotope dilution methods by GC/MS or LC/MS for pain prescription monitoring, clinical toxicology, urine drug testing or forensic analysis. Buprenorphine, sold under trade names such as Buprenex™, Subutex™ and Suboxone™, is an opiate drug used to treat opioid addiction and to moderate acute pain.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 1.0 mg/mL in methanol
Linear Formula C25H32D3NO4
MDL Number MFCD01861544
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 416.57

CERILLIANT is a registered trademark of Cerilliant Corporation