Learn More
CERILLIANT™ Naloxone-D5 Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore N1151ML
Description
A stable-labeled internal standard suitable for numerous quantitative LC/MS and GC/MS applications in forensic analysis, clinical toxicology testing, pharmaceutical analysis and isotope dilution methods. Naloxone is an opiate drug used to counter the effects of opioid overdose.Specifications
| 67-56-1 , 1261079-38-2 | |
| MFCD08063611 | |
| 1.0 mg/mL in methanol | |
| Ampule |
| C19H16NO4D5 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| 332.41 | |
| 1 mL |
CERILLIANT is a registered trademark of Cerilliant Corporation