Learn More
CERILLIANT™ N-Desmethylmirtazapine Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore D0861ML
Description
N-Desmethylmirtazapine is the major urinary metabolite of the tetracyclic antidepressant mirtazapine, marketed under the trade names Remeron™, Avanza, or Zispin for treatment of depression. The metabolite, also known as normirtazapine, is pharmacologically active. This Certified Spiking Solution™ is suitable for LC/MS or GC/MS methods in mirtazapine testing for forensic analysis, clinical toxicology, or urine drug testing.Specifications
| 75-05-8,61337-68-6 | |
| UN 1648 3 / PGII | |
| 251.33 | |
| 1 mL |
| C16H17N3 | |
| 1.0 mg/mL in acetonitrile | |
| Ampule | |
| 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation