missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ N-Desmethylclobazam Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Description
A Certified Solution Standard for use in LC or GC applications from forensic and forensic toxicology analysis to urine drug testing. N-Desmethylclobazam, found in urine, serum, and plasma, is the pharmacologically active, major metabolite of clobazam.
Specifications
Specifications
| CAS | 75-05-8, 22316-55-8 |
| Linear Formula | C15H11ClN2O2 |
| MDL Number | MFCD00871860 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 100 μg/mL in acetonitrile |
| Formula Weight | 286.71 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation