missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ N-Desmethylcitalopram Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore D0471ML
Description
N-Desmethylcitalopram is an active metabolite of the SSRI antidepressant drug citalopram. Citalopram, marketed as Celexa™ in the US and Canada, is used to treat major depression associated with mood disorders.Specifications
| 67-56-1,144025-14-9 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| N-Desmethylcitalopram hydrochloride solution | |
| Ampule | |
| 9.7°C (49.5°F) - closed cup |
| C19H19FN2O · HCl | |
| 1.0 mg/mL in methanol (as free base) | |
| 346.83 | |
| 1 mL |
CERILLIANT is a registered trademark of Cerilliant Corporation