missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ N-Desmethyl-cis-tramadol Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001162
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001162 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001162 Supplier Merck Emd Millipore Supplier No. D0231ML

Certified reference material

N-Desmethyl-cis-tramadol is a primary urinary and plasma metabolite of the synthetic analgesic tramadol. Tramadol, sold under trade names Ultram™, Conzip™, and Ryzolt™, relieves moderate to severe pain. This analytical reference standard is applicable for numerous LC/MS or GC/MS applications from pain prescription monitoring, urine drug testing and forensic analysis to clinical toxicology or pharmaceutical testing.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 1.0 mg/mL in methanol (as free base)
Linear Formula C15H24ClNO2
MDL Number MFCD18374605
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 285.81

CERILLIANT is a registered trademark of Cerilliant Corporation