missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ N-Desmethyl-cis-tramadol-D3 Hydrochloride Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001222
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001222 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001222 Supplier Merck Emd Millipore Supplier No. D1101ML

Certified reference material

N-Desmethyl-cis-tramadol is a primary urinary and plasma metabolite of the synthetic analgesic tramadol. Tramadol, sold under trade names Ultram, Conzip, and Ryzolt, relieves moderate to severe pain. This stable-labeled internal standard is suitable for the quantitation of N-Desmethyl-cis-tramadol in biological samples using GC/MS or LC/MS with numerous testing applications including forensic or clinical toxicology analysis, urine drug testing, or isotope dilution methods.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol (as free base)
Linear Formula C15H20D3NO2 · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 288.83

CERILLIANT is a registered trademark of Cerilliant Corporation