missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Mycophenolic acid Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Description
Assayed in therapeutic drug monitoring to ensure patients remain within the drug's therapeutic range, mycophenolic acid is an immunosuppressant drug and the active metabolite of the prodrug mycophenolate mofetil. This analytical standard is suitable for LC-MS/MS applications including therapeutic drug monitoring and other clinical or diagnostic applications.
Specifications
Specifications
| CAS | 75-05-8, 24280-93-1 |
| Linear Formula | C17H20O6 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 1.0 mg/mL in acetonitrile |
| Formula Weight | 320.34 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation