missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Mycophenolic acid-d3 Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001658
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001658 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001658 Supplier Merck Emd Millipore Supplier No. M1801ML
Only null left

Certified reference material

Assayed in therapeutic drug monitoring to ensure patients remain within the drug's therapeutic range, mycophenolic acid is an immunosuppressant drug and the active metabolite of the prodrug mycophenolate mofetil. This internal standard is suitable for quantification of mycophenolic acid levels in whole blood or plasma by LC-MS/MS for therapeutic drug monitoring and other clinical or diagnostic applications.

Specifications

CAS 75-05-8, 1185242-90-3
Linear Formula C17H17O6D3
MDL Number MFCD10001681
UN Number UN 1648 3 / PGII
Concentration 1.0 mg/mL in acetonitrile
Formula Weight 323.36
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation