missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Morphine Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001565
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001565 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001565 Supplier Merck Emd Millipore Supplier No. M0051ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified reference material

Morphine is a very potent opiate analgesic drug used to relieve both acute and chronic severe pain. Sold under trade names including MS Contin™, Kadian™, Oramorph™, and Roxanol, morphine is also a commonly abused recreational drug due to its widespread availability and high potential for addiction. This certified analytical reference standard is suitable for a variety of GC/MS or LC/MS applications from pain prescription monitoring, clinical toxicology and urine drug testing to pharmaceutical research and forensic analysis.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 1.0 mg/mL in methanol
Linear Formula C17H19NO3
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 285.34

CERILLIANT is a registered trademark of Cerilliant Corporation