missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Morphine-6-beta-D-glucuronide Solution, 100 μg/mL in Methanol:Water (1:1), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001614
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001614 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001614 Supplier Merck Emd Millipore Supplier No. M0961ML
Only null left

Certified reference material

Morphine-6-β-D-glucuronide is a primary plasma and urinary metabolite of the opiate analgesic morphine. Sold under trade names including MS Contin™, Kadian™, Oramorph™, and Roxanol for pain relief, morphine is also a commonly abused recreational drug. This certified analytical reference standard is suitable for a variety of GC/MS or LC/MS applications from pain prescription monitoring and urine drug testing to clinical toxicology and forensic analysis.

Specifications

CAS 67-56-1, 20290-09-9
Linear Formula C23H27NO9
MDL Number MFCD00079434
UN Number UN 1993C 3 / PGIII
Concentration 100 μg/mL in methanol:water (1:1)
Formula Weight 461.46
Packaging Ampule
Quantity 1 mL
Flash Point 24°C (75.2°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation