missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Modafinil-d10 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001616
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001616 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001616 Supplier Merck Emd Millipore Supplier No. M1031ML

Certified reference material

A new internal standard suitable for numerous GC/MS and LC/MS applications from clinical toxicology testing and forensic analysis to urine drug testing and isotope dilution methods. Modafinil is an analeptic drug sold under the brand names Provigil™ or Modavigil™ and approved by the US FDA for the treatment of narcolepsy, shift work sleep disorder, and excessive daytime sleepiness. A well known nootropic, the drug is often used as a performance-enhancer in academics and sports.

Specifications

CAS 75-05-8, 1219804-30-4
Linear Formula C15H5NO2SD10
MDL Number MFCD16039488
UN Number UN 1648 3 / PGII
Concentration 100 μg/mL in acetonitrile
Formula Weight 283.41
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation