missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Mitragynine Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Description
Mitragynine, commonly known as Kratom, is the most abundant alkaloid in the plant Mitragynine speciosa. Sold over the internet as herbal plant extracts, Kratom is noted for its psychoactive, stimulant, and sedative effects depending on the dosage level. According to DEA's Drugs and Chemicals of Concern page, mitragynine use can lead to addiction with adverse effects ranging from anorexia and weight loss to hallucinations and delusion.
Specifications
Specifications
| CAS | 67-56-1, 4098-40-2 |
| Linear Formula | C23H30N2O4 |
| MDL Number | MFCD09838906 |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Concentration | 100 μg/mL in methanol |
| Formula Weight | 398.50 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation