missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Methylphenidate-d9 Hydrochloride Solution, , Sold by MilliporeSigma™ Supelco™
Description
Methylphenidate is a psychostimulant prescribed for the treatment of attention-deficit hyperactivity disorder (ADHD), tachycardia and narcolepsy. This internal standard is suitable for quantitation of methylphenidate levels in urine, serum or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology or forensic analysis. Methylphenidate is sold under trade names Concerta™ and Ritalin™.
Specifications
Specifications
| CAS | 67-56-1, 1219804-02-0 |
| Linear Formula | C15H14NO2D9 |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Formula Weight | 258.40 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation