missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Methylone Hydrochloride, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Description
A synthetic stimulant of the amphetamine and cathinone classes also known as bk-MDMA or 3,4-methylenedioxy-N-methylcathinone. Significant abuse of this designer drug has been reported in the UK, Netherlands, Sweden, and New Zealand.
Specifications
Specifications
| Packaging | Ampule |
| CAS | 67-56-1 |
| Flash Point | 9.7°C (49.5°F) - closed cup |
| Concentration | 1.0 mg/mL in methanol (as free base) |
| Linear Formula | C11H13NO3 · HCl |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Synonym | Methylone hydrochloride solution |
| Quantity | 1 mL |
| Formula Weight | 243.69 |
CERILLIANT is a registered trademark of Cerilliant Corporation