missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Methoxyacetyl fentanyl Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001673
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001673 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001673 Supplier Merck Emd Millipore Supplier No. M2000.5ML

Certified reference material

Methoxyacetyl fentanyl is a designer drug and fentanyl analog that has been associated with numerous overdose deaths around the world. A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls for methoxyacetyl fentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing or forensic analysis applications. This certified solution standard is DEA-exempt, allowing laboratories to place orders without additional regulatory paperwork.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol (as free base)
Linear Formula C22H28N2O2 · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 0.5 mL
Formula Weight 388.93

CERILLIANT is a registered trademark of Cerilliant Corporation