missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Methocarbamol Solution, 1 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Description
Methocarbamol, sold under the brand name Robaxin, is a central muscle relaxant used to treat skeletal muscle spasms. This certified solution standard is applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from therapeutic drug monitoring to forensic analysis and clinical toxicology.
Specifications
Specifications
| CAS | 75-05-8, 532-03-6 |
| Linear Formula | C11H15NO5 |
| MDL Number | MFCD00057662 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 1 mg/mL in acetonitrile |
| Formula Weight | 241.24 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation