missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Methandienone Solution, 1.0 mg/mL, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001695
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001695 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001695 Supplier Merck Emd Millipore Supplier No. M9121ML

Certified reference material

A Certified Spiking Solution™ applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications such as sports testing, urine drug testing, clinical toxicology, and forensic analysis. Methandrostenolone, also called methandienone, is an anabolic steroid sold under the trade name Dianabol™. This performance enhancing drug has an extensive history of abuse by bodybuilders.

Specifications

CAS 110-71-4, 72-63-9
Linear Formula C20H28O2
MDL Number MFCD00003650
UN Number UN 2252 - class 3 - PG 2 - 1,2-Dimethoxyethane solution
Concentration 1.0 mg/mL
Formula Weight 300.44
Packaging Ampule
Quantity 1 mL
Flash Point -2°C (28.4°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation