missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ (+/-)-Methadone-D9 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001610
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001610 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001610 Supplier Merck Emd Millipore Supplier No. M0881ML

Certified reference material

A stable-labeled internal standard applicable for use in isotope dilution methods or quantitation of methadone levels by GC/MS or LC/MS in forensic analysis, urine drug testing, pain prescription monitoring or clinical toxicology applications. Methadone is a synthetic opioid used medically as an analgesic and as a maintenance anti-addictive for use in patients with opioid dependency. The drug is sold under several trade names such as Symoron, Dolophine™, Amidone, Methadose™, Physeptone, or Heptadon.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol
Linear Formula C21D9H18NO
MDL Number MFCD27966808
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 318.5

CERILLIANT is a registered trademark of Cerilliant Corporation