Learn More
CERILLIANT™ (+/-)-Methadone-D3 Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  Merck Emd Millipore M0211ML
Catalog No. 01001578
Only null left
Explore available promotions
Description
Description
A stable-labeled internal standard applicable for use in isotope dilution methods or quantitation of methadone levels by GC/MS or LC/MS in forensic analysis, urine drug testing, pain prescription monitoring or clinical toxicology applications. Methadone is a synthetic opioid used medically as an analgesic and as a maintenance anti-addictive for use in patients with opioid dependency. The drug is sold under several trade names such as Symoron, Dolophine™, Amidone, Methadose™, Physeptone, or Heptadon.
Specifications
Specifications
67-56-1,60263-63-0
MFCD00673285
1.0 mg/mL in methanol
312.46
1 mL
C21D3H24NO
UN 1230 - class 3 - PG 2 - Methanol, solution
DL-Methadone-d3 solution, DL-6-Dimethylamino-4,4-diphenyl-3-heptanone-1,1,1-d3 solution, DL-Methadone-(heptyl-1,1,1-d3) solution
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
CERILLIANT is a registered trademark of Cerilliant Corporation