missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Metformin Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001597
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001597 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001597 Supplier Merck Emd Millipore Supplier No. M0721ML
Only null left

Certified reference material

A Snap-N-Spike™ reference solution applicable to use in clinical toxicology by LC-MS/MS or GC/MS. Metformin is an antidiabetic drug which functions by suppressing liver production of glucose. This biguanide analog is used specifically for treatment of type 2 diabetes.

Specifications

CAS 67-56-1, 1115-70-4
Linear Formula C4H12ClN5
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 165.62
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation