Learn More
CERILLIANT™ Meprobamate Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore M0391ML
Description
Meprobamate is marketed as an anti-anxiety drug and muscle relaxant under the trade names Milltown, Meprospan, and Equanil. This Certified Spiking Solution™ is suitable for GC/MS or LC/MS testing applications in forensic analysis, clinical toxicology, urine drug testing, or pain prescription monitoring.Specifications
| 67-56-1,57-53-4 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| 218.25 | |
| 1 mL |
| C9H18N2O4 | |
| 1.0 mg/mL in methanol | |
| Ampule | |
| 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation