missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Meprobamate-D7 (2-methyl-1,3-propanediol-D7) Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001598
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001598 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001598 Supplier Merck Emd Millipore Supplier No. M0731ML
Only null left

Certified reference material

A stable-labeled internal standard suitable for meprobamate testing or isotope dilution methods by GC/MS or LC/MS in clinical toxicology, drug testing, pain prescription monitoring or forensic analysis. Marketed under the trade names Milltown, Meprospan, and Equanil, meprobamate is a minor tranquilizer used for the treatment of anxiety and muscle spasms.

Specifications

CAS 67-56-1
Linear Formula C9H11N2O4D7
MDL Number MFCD16294738
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 225.29
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation