missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Meprobamate-D3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Description
This internal standard is suitable for meprobamate testing or isotope dilution methods by LC/MS or GC/MS for clinical toxicology, forensic analysis, pain prescription monitoring, or urine drug testing applications. Marketed under the trade names Milltown, Meprospan, and Equanil, meprobamate is a minor tranquilizer used for the treatment of anxiety and muscle spasms.
Specifications
Specifications
| CAS | 67-56-1, 1185106-66-4 |
| Linear Formula | C9H15D3N2O4 |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Concentration | 100 μg/mL in methanol |
| Formula Weight | 221.27 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 9.7°C (49.5°F) |
CERILLIANT is a registered trademark of Cerilliant Corporation