missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Mephedrone Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001628
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001628 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001628 Supplier Merck Emd Millipore Supplier No. M1381ML

Certified reference material

Mephedrone, also known as 4-methylmethcathinone, is a synthetic stimulant of the amphetamine and cathinone classes. Significant abuse of this new recreational drug has been reported within the last year in the UK; Sweden; and Toronto, Canada with confiscation of the drug in states including Oregon.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 1.0 mg/mL in methanol (as free base)
Linear Formula C11H16ClNO
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 213.7

CERILLIANT is a registered trademark of Cerilliant Corporation