missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ MDAI Hydrochloride Solution, 100 μg/mL in Acetonitrile:Water (9:1) with 5% 1 M HCl, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001633
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001633 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001633 Supplier Merck Emd Millipore Supplier No. M1441ML

Certified reference material

MDAI, or 5,6-Methylenedioxy-2-aminoindane, is a new stimulant and club/designer drug with significant cases of abuse reported in Asia, Europe and the US. Sold over the Internet as a “bath salt,” MDAI is a potential successor to mephedrone as the next legal high drug.

Specifications

CAS 75-05-8, 132741-81-2
Linear Formula C10H11NO2 · HCl
UN Number UN 1648 3 / PGII
Concentration 100 μg/mL in acetonitrile:water (9:1) with 5% 1 M HCl (as free base)
Formula Weight 213.66
Packaging Ampule
Quantity 1 mL
Flash Point 13°C (55.4°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation