Learn More
CERILLIANT™ (+/-)-MDA Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  Merck Emd Millipore M0121ML
Catalog No. 01001571
Only null left
Explore available promotions
Description
Description
A certified solution standard suitable to use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug testing. MDA, or 3,4-methylenedioxyamphetamine, is an illicit recreational drug of the amphetamine and phenethylamine classes. Also known as tenamphetamine, MDA is abused for its psychedelic and hallucinogenic effects, similar to those of MDMA (Ecstasy).
Specifications
Specifications
67-56-1,4764-17-4
MFCD00867901
1.0 mg/mL in methanol
179.22
1 mL
C10H13NO2
UN 1230 - class 3 - PG 2 - Methanol, solution
(+/-)-3,4-Methylenedioxyamphetamine
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
CERILLIANT is a registered trademark of Cerilliant Corporation