missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ (+/-)-MBDB Hydrochloride, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Description
A certified solution standard suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, urine drug testing, or pharmaceutical research. MBDB is an illicit drug structurally related to MDMA, with similar effects including euphoria and mild stimulation.
Specifications
Specifications
| CAS | 67-56-1, 128767-12-4 |
| Linear Formula | C12H17NO2 · HCl |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Concentration | 1.0 mg/mL in methanol (as free base) |
| Synonym | (+\-)-MBDB hydrochloride solution |
| Formula Weight | 243.73 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation