missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ LSD Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Description
LSD, or lysergic acid diethylamide, is an illicit psychedelic drug with potent hallucinogenic effects such as altered thinking processes, closed- and open-eye visuals, synesthesia and an altered sense of time. This Snap-N-Spike™ reference solution is suitable for use in urine drug testing, clinical toxicology, or forensic analysis by LC-MS/MS or GC/MS.
Specifications
Specifications
| CAS | 75-05-8, 50-37-3 |
| Linear Formula | C20H25N3O |
| MDL Number | MFCD00135795 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 1.0 mg/mL in acetonitrile |
| Synonym | Lysergic acid diethylamide |
| Formula Weight | 323.43 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation