missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Levetiracetam-D6 Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001542
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001542 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001542 Supplier Merck Emd Millipore Supplier No. L0311ML
Only null left

Certified reference material

An internal standard suitable for levetiracetam GC/MS or LC/MS testing or isotope dilution methods in forensic analysis, clinical toxicology, pain prescription monitoring, or urine drug testing. Levetiracetam, sold under the trade name Keppra™, is an antiepileptic drug used primarily for treatment of seizures along with occasional use in treating neuropathic pain.

Specifications

CAS 67-56-1
Linear Formula C8D6H8N2O2
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Synonym Levetiracetam-D6 solution
Formula Weight 176.25
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation