missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Ketamine Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001521
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001521 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001521 Supplier Merck Emd Millipore Supplier No. K0021ML

Certified reference material

Marketed as Ketanest™, Ketaset™ and Ketalar™, ketamine is a widely used anesthetic, particularly in emergency and critical care medicine. The drug is also abused at clubs, raves, or parties for its noted hallucinogenic and dissociative properties and prescribed by some doctors as an off-label treatment for depression. This certified solution standard is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS testing applications from forensic analysis and clinical toxicology to urine drug testing.

Specifications

CAS 67-56-1, 1867-66-9
Linear Formula C13H17Cl2NO
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 274.19
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation