missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Isobutyryl fentanyl Hydrochloride Solution, 100 μg/mL in Methanol (as a free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001506
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001506 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001506 Supplier Merck Emd Millipore Supplier No. I0380.5ML

Certified reference material

A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls for isobutyryl fentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, prescription monitoring, or forensic analysis applications. Isobutyryl fentanyl is a fentanyl analog and sold on the illicit market as a designer drug or in combination with heroin.

Specifications

Packaging Ampule
CAS 67-56-1
Concentration 100 μg/mL in methanol (as a free base)
Linear Formula C23H30N2O · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 0.5 mL
Formula Weight 386.96

CERILLIANT is a registered trademark of Cerilliant Corporation