missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Insulin from bovine pancreas Solution, 100 μg/mL (PBS pH 7.2), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001503
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001503 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001503 Supplier Merck Emd Millipore Supplier No. I0350.5ML

Certified reference material

Bovine insulin differs from human insulin in three amino acid residues and is therefore used as internal standard for human insulin. This Certified Spiking Solution™ of fully-intact bovine insulin protein is suitable for quantification of human insulin in LC-MS/MS insulin testing methods. Insulin is a peptide hormone, composed of 51 amino acids, produced by beta cells of the pancreatic islets. Insulin regulates the metabolism of carbohydrates, fats and protein by promoting the absorption of, especially glucose from the blood into liver, fat and skeletal muscle cells. Insulin levels are routinely measured to evaluate insulin production; diagnose insulin-producing tumors; determine hypoglycemia or to identify insulin resistance.

Specifications

CAS 11070-73-8
Linear Formula C254H377N65O75S6
Concentration 100 μg/mL (PBS pH 7.2)
Formula Weight 5733.49
Packaging Ampule
Quantity 0.5 mL

CERILLIANT is a registered trademark of Cerilliant Corporation