missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ IGF-1 Solution, 100 μg/mL (2% Acetic acid, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001501
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001501 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001501 Supplier Merck Emd Millipore Supplier No. I0331ML

Certified reference material

Insulin-like growth factor 1 (IGF-1), also called somatomedin C, is a protein hormone that plays an important role in childhood growth and continues to have anabolic effects in adults. Measuring IGF-1 levels is used to test for growth hormone defficiency and excess in acromegaly and gigantism. This certified Snap-N-Spike™ solution of the fully-intact IGF-1 protein is suitable for use as starting material for calibrators and controls in IGF-1 LC/MS testing applications.

Specifications

CAS 64-19-7
Concentration 100 μg/mL (2% Acetic acid
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation