Learn More
CERILLIANT™ Ibuprofen Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  Merck Emd Millipore I0091ML
Catalog No. 01001485
Only null left
Explore available promotions
Description
Description
Ibuprofen is a non-steroidal anti-inflammatory drug (NSAID) used to treat fever, pain, and inflammation. Ibuprofen functions as an unspecific COX inhibitor. This Certified Spiking Solution™ is applicable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or pharmaceutical research.
Specifications
Specifications
67-56-1,15687-27-1
UN 1230 - class 3 - PG 2 - Methanol, solution
205.27
1 mL
C13H17O2
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
CERILLIANT is a registered trademark of Cerilliant Corporation